Learn More
Medchemexpress LLC ZD 7155 hydrochloride | 146709-78-6 | MFCD00921888; MFCD30342322 | 98.3% | 475.0 g/mol | C26H27ClN6O | 100 MG
SDP
Catalog No. 5000247207
Encompass_Preferred
ZD 7155 hydrochloride is an angiotensin II type 1 (AT1) receptor antagonist provided as a research-grade analytical standard for pharmacological and cardiovascular studies. It is used in receptor binding assays, mechanistic in vitro experiments, and in vivo pharmacodynamic investigations to probe AT1-mediated pathways.
  • Potent, selective AT1 receptor antagonist.
  • High purity suitable for analytical and research applications.
  • Available as solid and solution preparations for flexible use in assays.
  • Soluble in common organic solvents such as DMSO for assay preparation.
  • Suitable for in vitro receptor binding and in vivo pharmacodynamic studies.
Catalog No. 50-002-47207 Supplier Medchemexpress LLC Supplier No. HY102093100MG
ZD 7155 hydrochloride is an angiotensin II type 1 (AT1) receptor antagonist provided as a research-grade analytical standard for pharmacological and cardiovascular studies. It is used in receptor binding assays, mechanistic in vitro experiments, and in vivo pharmacodynamic investigations to probe AT1-mediated pathways.
  • Potent, selective AT1 receptor antagonist.
  • High purity suitable for analytical and research applications.
  • Available as solid and solution preparations for flexible use in assays.
  • Soluble in common organic solvents such as DMSO for assay preparation.
  • Suitable for in vitro receptor binding and in vivo pharmacodynamic studies.