Learn More
Cayman Chemical ZD 7155hydrochlorIde 1mg
SDP
Catalog No. 502914027
Encompass_Preferred
An AT1 receptor antagonist (IC50 = 3.3 nM); inhibits angiotensin II-induced pressor responses in conscious normotensive rats; decreases mean arterial pressure in conscious spontaneously hypertensive rats at 1.082 µmol/kg; improves motor nerve conduction velocity in the sciatic nerve in a rat model of streptozotocin-induced diabetic neuropathy at 10 mg/kg in the drinking water; decreases disease severity and NF-κB and AP-1 activation in a mouse model of LPS-induced acute lung injury at 10 mg/kg
Catalog No. 50-291-4027 Supplier Cayman Chemical Supplier No. 365761 MG
An AT1 receptor antagonist (IC50 = 3.3 nM); inhibits angiotensin II-induced pressor responses in conscious normotensive rats; decreases mean arterial pressure in conscious spontaneously hypertensive rats at 1.082 µmol/kg; improves motor nerve conduction velocity in the sciatic nerve in a rat model of streptozotocin-induced diabetic neuropathy at 10 mg/kg in the drinking water; decreases disease severity and NF-κB and AP-1 activation in a mouse model of LPS-induced acute lung injury at 10 mg/kg