Learn More
Apexbio Technology LLC ZD 7288(Synonyms: ZD7288, Zatebradine hydrochloride, HCN channel inhibitor ZD7288), 10mg, CAS: 133059-99-1.
SDP
Catalog No. 5000060050
Encompass_Preferred
ZD 7288 (CAS 133059-99-1) is a selective inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels These channels mediate Na and K currents notably the hyperpolarization-activated current (I h) contributing to neuronal excitability and rhythmic activity ZD 7288 reduces I h in a concentration-dependent voltage- and time-sensitive manner as demonstrated in rat facial motoneurons (IC 50 0 2 M) and hippocampal CA1 pyramidal neurons Additionally it inhibits T-type calcium currents in rat CA1 pyramidal cells ZD 7288 is utilized in research investigating neuronal excitability mechanisms pain signaling and chronic visceral pain models
Catalog No. 50-000-60050 Supplier Apexbio Technology LLC Supplier No. B654210
ZD 7288 (CAS 133059-99-1) is a selective inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels These channels mediate Na and K currents notably the hyperpolarization-activated current (I h) contributing to neuronal excitability and rhythmic activity ZD 7288 reduces I h in a concentration-dependent voltage- and time-sensitive manner as demonstrated in rat facial motoneurons (IC 50 0 2 M) and hippocampal CA1 pyramidal neurons Additionally it inhibits T-type calcium currents in rat CA1 pyramidal cells ZD 7288 is utilized in research investigating neuronal excitability mechanisms pain signaling and chronic visceral pain models