Learn More
Apexbio Technology LLC ZD 9379 10mg
SDP
Catalog No. 5000088903
Encompass_Preferred
ZD 9379 (CAS 170142-20-8) is a small molecule that acts as an antagonist at the glycine-binding site of the NMDA receptor Exhibiting oral bioavailability and capable of crossing the blood-brain barrier ZD 9379 modulates neuronal signaling by inhibiting NMDA receptor-mediated pathways Experimental studies have demonstrated its capacity to suppress functions associated with the central nervous system Due to its pharmacological profile ZD 9379 is primarily employed in the investigation of neurological disease models and the elucidation of NMDA receptor-related mechanisms
Catalog No. 50-000-88903 Supplier Apexbio Technology LLC Supplier No. B540910
ZD 9379 (CAS 170142-20-8) is a small molecule that acts as an antagonist at the glycine-binding site of the NMDA receptor Exhibiting oral bioavailability and capable of crossing the blood-brain barrier ZD 9379 modulates neuronal signaling by inhibiting NMDA receptor-mediated pathways Experimental studies have demonstrated its capacity to suppress functions associated with the central nervous system Due to its pharmacological profile ZD 9379 is primarily employed in the investigation of neurological disease models and the elucidation of NMDA receptor-related mechanisms