Learn More
Medchemexpress LLC HY-101346 10mg Medchemexpress, ZD7288 CAS:133059-99-1 Purity:>98%
SDP
Catalog No. 501967830
Encompass_Preferred
Medchemexpress, HY-101346 10mg ZD7288 CAS:133059-99-1 ZD7288 is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7830 Supplier Medchemexpress LLC Supplier No. HY10134610MG
Medchemexpress, HY-101346 10mg ZD7288 CAS:133059-99-1 ZD7288 is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.