Learn More
Abcam ZD7288, Sino-atrial node function modulator, 10MG
Catalog No. 502584695
Encompass_Preferred
MW 292.81 Da, Purity >98%. Sino-atrial node function modulator; blocks the hyperpolarisation activated inward current If. Also inhibits Ih in central neurons and inhibits synaptic transmission.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4695 Supplier Abcam Supplier No. AB12010210MG
Add to Cart
Add to Cart
MW 292.81 Da, Purity >98%. Sino-atrial node function modulator; blocks the hyperpolarisation activated inward current If. Also inhibits Ih in central neurons and inhibits synaptic transmission.
The product is subject to the following: Abcam Restricted Use Statement