Learn More
Cayman Chemical Z-D-Arg-Gly-Arg-pNA (hydrochloride), Purity: ≥98%, CAS: 113711-77-6, 10 mg
SDP
Catalog No. 502939715
Encompass_Preferred
A colorimetric Factor Xa substrate; cleavage of the RGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity
Catalog No. 50-293-9715 Supplier Cayman Chemical Supplier No. 2758310 MG
A colorimetric Factor Xa substrate; cleavage of the RGR peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of Factor Xa activity