Learn More
Aviva Systems Biology Corp Zdhhc11 Antibody - middle region (ARP49871_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zdhhc11. It was validated on Western Blot. . Alternate names: Zdhhc20, Znf399ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-458