Learn More
Aviva Systems Biology Corp ZDHHC13 Antibody - N-terminal region (ARP44398_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44398_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZDHHC13. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZDHHC13. Alternate names: FLJ10852, FLJ10941, HIP14L, HIP3RP, MGC64994ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58799