Learn More
Aviva Systems Biology Corp ZDHHC17 Antibody - middle region (ARP47142_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47142_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZDHHC17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZDHHC17. Alternate names: HIP14, HIP3, HSPC294, HYPH, KIAA0946ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59471