Learn More
Aviva Systems Biology Corp ZDHHC18 Antibody - middle region (ARP42673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZDHHC18. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZDHHC18. Alternate names: DKFZp667O2416, DHHC18, DHHC-18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-018