Learn More
Aviva Systems Biology Corp ZDHHC19 Antibody - middle region (ARP44883_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44883_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZDHHC19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZDHHC19. Alternate names: DHHC19ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58928