Learn More
Aviva Systems Biology Corp ZDHHC21 Antibody - middle region (ARP55783_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55783_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZDHHC21. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZDHHC21. Alternate names: DNZ1, DHHC-21, HSPC097, 9130404H11RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-079