Learn More
Aviva Systems Biology Corp ZDHHC3 Antibody - C-terminal region (ARP59576_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59576_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZDHHC3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ZDHHC3. Alternate names: DKFZp313D2314, FLJ20209, FLJ45940, GODZ, ZNF373, DHHC-3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-746