Learn More
Bio Rad Laboratories ZE-Series QC Beads, 5 mL, 3/pk
Catalog No. NC1567169
Encompass
Includes 3–3.4 um single-peak beads containing dyes that emit in all fluorescence channels from ultraviolet to far red. Designed for checking optical alignment in all channels of the ZE5 Cell Analyzer.
Catalog No. NC1567169 Supplier Bio Rad Laboratories Supplier No. 12004403
Includes 3–3.4 um single-peak beads containing dyes that emit in all fluorescence channels from ultraviolet to far red. Designed for checking optical alignment in all channels of the ZE5 Cell Analyzer.