Learn More
Medchemexpress LLC β-Zearalanol-d4 | 1778735-09-3 | 1 MG
SDP
Catalog No. 5000417778
Encompass_Preferred
β-Zearalanol-d4 is a deuterium-labeled Beta-Zearalanol, a mycotoxin produced by Fusarium spp. This compound is known to induce apoptosis and oxidative stress in mammalian reproductive cells and is a derivative of zearalenone (ZEA). Deuterated compounds like β-Zearalanol-d4 are valuable as tracers and internal standards in drug development and analytical research, as their stable heavy isotopes can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
  • For research use only
Catalog No. 50-004-17778 Supplier Medchemexpress LLC Supplier No. HYN6740S1MG
β-Zearalanol-d4 is a deuterium-labeled Beta-Zearalanol, a mycotoxin produced by Fusarium spp. This compound is known to induce apoptosis and oxidative stress in mammalian reproductive cells and is a derivative of zearalenone (ZEA). Deuterated compounds like β-Zearalanol-d4 are valuable as tracers and internal standards in drug development and analytical research, as their stable heavy isotopes can influence pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
  • For research use only