Learn More
Enzo Life Sciences Zearalenone (10mg). CAS: 17924-92-4
SDP
Supplier:  Enzo Life Sciences ALX630105M010
Encompass_Preferred
Zearalenone (ZEN) is an estrogenic mycotoxin in animals and a phytohormone in plants. Inducer of sister chromatid exchange and chromosomal aberration. Acts as a protonophoric uncoupler in plant mitochondria. Alternative name: Benzoxacyclotetradecin-1,7(8H)-dione, ZEN. Purity: ≥99% (HPLC, TLC). Solubility: Soluble in methanol  (5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9629