Learn More
Medchemexpress LLC HY-103447 10mg Medchemexpress, Zearalenone CAS:17924-92-4 Purity:>98%
SDP
Catalog No. 501968108
Encompass_Preferred
Medchemexpress, HY-103447 10mg Zearalenone CAS:17924-92-4 Zearalenone is a mycotoxin produced mainly by fungi belonging to the genus Fusarium in foods and feeds. Possess oestrogenic activity in pigs, cattle and sheep, with low acute toxicity. Causes precocious development of mammae and other estrogenic effects in young gilts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8108 Supplier Medchemexpress LLC Supplier No. HY10344710MG
Medchemexpress, HY-103447 10mg Zearalenone CAS:17924-92-4 Zearalenone is a mycotoxin produced mainly by fungi belonging to the genus Fusarium in foods and feeds. Possess oestrogenic activity in pigs, cattle and sheep, with low acute toxicity. Causes precocious development of mammae and other estrogenic effects in young gilts. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.