Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zearalenone | 17924-92-4 | MFCD00133085 | 98.9% | 318.36 g/mol | C18H22O5 | 25 MG
SDP

Catalog No. 5000244857
Encompass_Preferred

Zearalenone is a nonsteroidal estrogenic mycotoxin provided as an analytical standard for research and analytical applications, including method development, calibration, and mycotoxin screening.

  • Purity: 98.9%.
  • CAS number: 17924-92-4.
  • Molecular formula: C18H22O5; molecular weight: 318.36 g/mol.
  • Appearance: white to off-white solid.
  • Pack size: 25 mg vial.
  • Storage: store under nitrogen; in solvent, -80°C (1 year) or -20°C (6 months).
  • Applications: analytical standard for mycotoxin detection, method development, and calibration.

Catalog No. 50-002-44857 Supplier Medchemexpress LLC Supplier No. HY10344725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zearalenone is a nonsteroidal estrogenic mycotoxin provided as an analytical standard for research and analytical applications, including method development, calibration, and mycotoxin screening.

  • Purity: 98.9%.
  • CAS number: 17924-92-4.
  • Molecular formula: C18H22O5; molecular weight: 318.36 g/mol.
  • Appearance: white to off-white solid.
  • Pack size: 25 mg vial.
  • Storage: store under nitrogen; in solvent, -80°C (1 year) or -20°C (6 months).
  • Applications: analytical standard for mycotoxin detection, method development, and calibration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.