Learn More
Apexbio Technology LLC Zearalenone(Synonyms: F-2 toxin, Mycotoxin F2, Zearalenolactone, RAL, ZEA, Zenon), 50mg, CAS: 17924-92-4.
SDP
Catalog No. 5000060253
Encompass_Preferred
Zearalenone (CAS 17924-92-4) is a mycotoxin produced primarily by Fusarium fungi frequently contaminating feed and food products It exhibits estrogenic activity through interaction with estrogen receptors thus altering endocrine signaling pathways Although acutely low in toxicity zearalenone effectively modulates estrogen receptor-mediated responses influencing hormonal balance in livestock species such as pigs cattle and sheep In biomedical research zearalenone serves as a tool compound to study estrogen receptor-mediated mechanisms and endocrine disruption particularly in young animals within veterinary pharmacological contexts
Catalog No. 50-000-60253 Supplier Apexbio Technology LLC Supplier No. B755650
Zearalenone (CAS 17924-92-4) is a mycotoxin produced primarily by Fusarium fungi frequently contaminating feed and food products It exhibits estrogenic activity through interaction with estrogen receptors thus altering endocrine signaling pathways Although acutely low in toxicity zearalenone effectively modulates estrogen receptor-mediated responses influencing hormonal balance in livestock species such as pigs cattle and sheep In biomedical research zearalenone serves as a tool compound to study estrogen receptor-mediated mechanisms and endocrine disruption particularly in young animals within veterinary pharmacological contexts