Learn More
Medchemexpress LLC Zearalenone (standard) | 17924-92-4 | MFCD00133085 | 98.9% | 318.36 g/mol | C18H22O5 | 1 ML
SDP
Catalog No. 5000245136
Encompass_Preferred
Zearalenone is a fungal mycotoxin supplied as a ready-to-use 10 mM solution in DMSO for research and analytical assays. The solution offers consistent concentration for assay setup, and is provided at high chemical purity for reliable results.
  • Ready-to-use 10 mM solution in DMSO
  • Convenient 1 mL vial for small-scale assays
  • High purity for analytical reproducibility
  • Accurate concentration to simplify assay setup
  • Intended for research and analytical applications only
Catalog No. 50-002-45136 Supplier Medchemexpress LLC Supplier No. HY10344710MM 1ML
Zearalenone is a fungal mycotoxin supplied as a ready-to-use 10 mM solution in DMSO for research and analytical assays. The solution offers consistent concentration for assay setup, and is provided at high chemical purity for reliable results.
  • Ready-to-use 10 mM solution in DMSO
  • Convenient 1 mL vial for small-scale assays
  • High purity for analytical reproducibility
  • Accurate concentration to simplify assay setup
  • Intended for research and analytical applications only