Learn More
Medchemexpress LLC Zearalenone (standard) | 17924-92-4 | MFCD00133085 | 98.9% | 318.36 g/mol | C18H22O5 | 25 MG
SDP
Catalog No. 5000246683
Encompass_Preferred
Zearalenone (standard) is an analytical reference standard of the mycotoxin zearalenone (CAS 17924-92-4) supplied as a solid for analytical and research use. It is intended for qualitative and quantitative method development and testing by HPLC, GC, and MS, and is supplied at high purity for reliable analytical performance.
  • Analytical standard for HPLC, GC, and MS.
  • High purity (≈98.9%) for reliable quantitation.
  • Solid form for easy storage and handling.
  • Suitable for qualitative and quantitative method development.
  • Includes certificate of analysis for quality verification.
Catalog No. 50-002-46683 Supplier Medchemexpress LLC Supplier No. HY103447R25MG
Zearalenone (standard) is an analytical reference standard of the mycotoxin zearalenone (CAS 17924-92-4) supplied as a solid for analytical and research use. It is intended for qualitative and quantitative method development and testing by HPLC, GC, and MS, and is supplied at high purity for reliable analytical performance.
  • Analytical standard for HPLC, GC, and MS.
  • High purity (≈98.9%) for reliable quantitation.
  • Solid form for easy storage and handling.
  • Suitable for qualitative and quantitative method development.
  • Includes certificate of analysis for quality verification.