Learn More
Research Products International Corp Zeatin, Trans Isomer [6-(4-Hydroxy-3-methylbut-2-enylamino)purine)], 100 Milligrams
SDP
Catalog No. 50213743
Encompass_Preferred
Naturally occurring cytokinin growth regulator. Promotes cell division and also influences other steps of the cell cycle. High concentrations produce advantitious shoot formation. Other uses include prevention of leaf senescence, reversal of toxin affects on shoots and inhibiting excessive root formation. Off white to yellow crystals. Plant cell culture tested.
Catalog No. 50-213-743 Supplier Research Products International Corp Supplier No. Z114000.1
Naturally occurring cytokinin growth regulator. Promotes cell division and also influences other steps of the cell cycle. High concentrations produce advantitious shoot formation. Other uses include prevention of leaf senescence, reversal of toxin affects on shoots and inhibiting excessive root formation. Off white to yellow crystals. Plant cell culture tested.