Learn More
Bioworld Zeatin, 1 g
Supplier:  Bioworld 306310194
Encompass_Preferred
ZeatinSynonyms: 6-(4-Hydroxy-3-methylbut-2-enylamino)purine; N6-(4-Hydroxy-3-methyl-2-buten-1-yl)adenine Cytokinin growth regulator Promotes cell division High concentrations produce advantitious shoot formationOther uses include prevention of leaf senescence, reversal of toxin affects on shoots, and inhibiting excessive root formationPlant cell culture testedHigh Purity GradeSoluble in 1M NaOHSolubilizeat 1 mg/mLin 0.1N KOH
Catalog No. 50-255-077