Learn More
Abcam Zeatin, cytokinin growth regulator, 100MG
Catalog No. 502704592
Encompass_Preferred
MW 219.2 g/mol, Purity >98%. Coconut milk-derived plant hormone (cytokinin) that promotes growth of lateral buds, stimulates cell division (Lateral Dominance), callus initiation and seed germination. Promotes multiple shoot generation from internodes and high frequency thick spread roots.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4592 Supplier Abcam Supplier No. AB146530100MG
MW 219.2 g/mol, Purity >98%. Coconut milk-derived plant hormone (cytokinin) that promotes growth of lateral buds, stimulates cell division (Lateral Dominance), callus initiation and seed germination. Promotes multiple shoot generation from internodes and high frequency thick spread roots.
The product is subject to the following: Abcam Restricted Use Statement