Learn More
Abcam Zeatin, cytokinin growth regulator, 1G
Catalog No. 502704591
Encompass_Preferred
MW 219.2 g/mol, Purity >98%. Coconut milk-derived plant hormone (cytokinin) that promotes growth of lateral buds, stimulates cell division (Lateral Dominance), callus initiation and seed germination. Promotes multiple shoot generation from internodes and high frequency thick spread roots.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4591 Supplier Abcam Supplier No. AB1465301G
MW 219.2 g/mol, Purity >98%. Coconut milk-derived plant hormone (cytokinin) that promotes growth of lateral buds, stimulates cell division (Lateral Dominance), callus initiation and seed germination. Promotes multiple shoot generation from internodes and high frequency thick spread roots.
The product is subject to the following: Abcam Restricted Use Statement