Learn More
Research Products International Corp Zeatin Riboside [9-(B-D-Ribofuranosyl)-Trans-Zeatin], 50 Milligrams
SDP
Catalog No. 50213747
Encompass_Preferred
Naturally occurring cytokinin riboside. Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Chromatographically purified and recrystallized. Plant cell culture tested.
Mixed isomers: trans-isomer 97%, cis-isomer >2%.
Catalog No. 50-213-747 Supplier Research Products International Corp Supplier No. Z114020.05
Naturally occurring cytokinin riboside. Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Chromatographically purified and recrystallized. Plant cell culture tested.
Mixed isomers: trans-isomer 97%, cis-isomer >2%.