Learn More
Bioworld Trans Zeatin Riboside, 50 mg
Catalog No. 50255079
Encompass_Preferred
Zeatin ribosideSynonyms: 9-(β-D-Ribofuranosyl)-trans-zeatin; N6-(trans-4-Hydroxy-3-methyl-2-buten-1-yl)adenosineCytokinin ribosideControl of shoot growth and formation, cell division and inhibition of leaf senescencePlant cell culture testedAppearance: white to yellow-white powderMelting Point: 176 °CSoluble in water
Catalog No. 50-255-079 Supplier Bioworld Supplier No. 306310202
Zeatin ribosideSynonyms: 9-(β-D-Ribofuranosyl)-trans-zeatin; N6-(trans-4-Hydroxy-3-methyl-2-buten-1-yl)adenosineCytokinin ribosideControl of shoot growth and formation, cell division and inhibition of leaf senescencePlant cell culture testedAppearance: white to yellow-white powderMelting Point: 176 °CSoluble in water