Learn More
Alkali Scientific Zeatin Riboside [9 (B D Ribofuranosyl) Trans Zeatin], 1 Mg
SDP
Catalog No. 5000661576
Encompass_Preferred
Zeatin Riboside (Trans Zeatin), 1 mg, is a cytokinin widely used in plant tissue culture to induce cell division and stimulate shoot formation. This reagent is often used in small-scale studies and experiments aimed at understanding plant growth regulation. Zeatin riboside works by enhancing cell division and delaying the onset of senescence, which allows for better growth and proliferation of plant tissues in culture. This 1mg pack is ideal for researchers and laboratories requiring a small quantity of reagent for tissue culture applications, plant regeneration experiments, and genetic modification studies. It supports the efficient cultivation of healthy plantlets in vitro.
Catalog No. 50-006-61576 Supplier Alkali Scientific Supplier No. ZEA006CP
Zeatin Riboside (Trans Zeatin), 1 mg, is a cytokinin widely used in plant tissue culture to induce cell division and stimulate shoot formation. This reagent is often used in small-scale studies and experiments aimed at understanding plant growth regulation. Zeatin riboside works by enhancing cell division and delaying the onset of senescence, which allows for better growth and proliferation of plant tissues in culture. This 1mg pack is ideal for researchers and laboratories requiring a small quantity of reagent for tissue culture applications, plant regeneration experiments, and genetic modification studies. It supports the efficient cultivation of healthy plantlets in vitro.