Learn More
Alkali Scientific Zeatin Riboside [9 (B D Ribofuranosyl) Trans Zeatin], 25 Mg
SDP
Catalog No. 5000661789
Encompass_Preferred
Zeatin Riboside (Trans Zeatin), 25 mg, is a naturally occurring cytokinin used in plant tissue culture for promoting cell division and differentiation. This reagent is particularly important for inducing shoot formation and enhancing the growth of plant cells in vitro. Zeatin riboside is a ribosylated form of zeatin, which is more stable and effective in stimulating plant growth and morphogenesis. This 25mg pack is ideal for small-scale tissue culture projects and is commonly used in plant regeneration studies, genetic modification experiments, and the production of genetically engineered plants. Zeatin riboside helps researchers optimize tissue culture conditions for better plant growth and productivity.
Catalog No. 50-006-61789 Supplier Alkali Scientific Supplier No. ZEA004CP
Zeatin Riboside (Trans Zeatin), 25 mg, is a naturally occurring cytokinin used in plant tissue culture for promoting cell division and differentiation. This reagent is particularly important for inducing shoot formation and enhancing the growth of plant cells in vitro. Zeatin riboside is a ribosylated form of zeatin, which is more stable and effective in stimulating plant growth and morphogenesis. This 25mg pack is ideal for small-scale tissue culture projects and is commonly used in plant regeneration studies, genetic modification experiments, and the production of genetically engineered plants. Zeatin riboside helps researchers optimize tissue culture conditions for better plant growth and productivity.