Learn More
Alkali Scientific Zeatin Riboside [9 (B D Ribofuranosyl) Trans Zeatin], 250 Mg
SDP
Catalog No. 5000661790
Encompass_Preferred
Zeatin Riboside (Trans Zeatin), 250 mg, is a cytokinin used to stimulate cell division, promote shoot formation, and prevent senescence in plant tissue cultures. This compound is essential for optimizing plant regeneration protocols, and its role in promoting morphogenesis makes it invaluable in plant biotechnology. The ribosylated form of zeatin is more stable, enhancing its effectiveness in plant tissue culture applications. This 250mg pack provides ample material for large-scale experiments and research on plant growth regulation. Zeatin riboside is frequently used in genetic transformation studies, the development of genetically modified crops, and in improving agricultural productivity through tissue culture techniques.
Catalog No. 50-006-61790 Supplier Alkali Scientific Supplier No. ZEA005CP
Zeatin Riboside (Trans Zeatin), 250 mg, is a cytokinin used to stimulate cell division, promote shoot formation, and prevent senescence in plant tissue cultures. This compound is essential for optimizing plant regeneration protocols, and its role in promoting morphogenesis makes it invaluable in plant biotechnology. The ribosylated form of zeatin is more stable, enhancing its effectiveness in plant tissue culture applications. This 250mg pack provides ample material for large-scale experiments and research on plant growth regulation. Zeatin riboside is frequently used in genetic transformation studies, the development of genetically modified crops, and in improving agricultural productivity through tissue culture techniques.