Learn More
Alkali Scientific Zeatin Riboside, [9 (B D Ribofuranosyl) Trans Zeatin], 50 Mg
SDP
Catalog No. 5000661791
Encompass_Preferred
Zeatin Riboside is a plant growth regulator in powder form, commonly used in laboratory applications to promote cell division and growth in plants. It is a cytokinin, a class of plant hormones that help regulate cell cycle progression, and it plays a vital role in shoot initiation, leaf development, and overall plant vigor. Zeatin Riboside is utilized in plant biotechnology to enhance tissue culture procedures, stimulate bud formation, and improve regeneration rates in various plant species. It is an essential reagent for research on plant development and plant tissue culture, and is used in agricultural research to optimize plant growth under controlled conditions.
Catalog No. 50-006-61791 Supplier Alkali Scientific Supplier No. ZEA001CP
Zeatin Riboside is a plant growth regulator in powder form, commonly used in laboratory applications to promote cell division and growth in plants. It is a cytokinin, a class of plant hormones that help regulate cell cycle progression, and it plays a vital role in shoot initiation, leaf development, and overall plant vigor. Zeatin Riboside is utilized in plant biotechnology to enhance tissue culture procedures, stimulate bud formation, and improve regeneration rates in various plant species. It is an essential reagent for research on plant development and plant tissue culture, and is used in agricultural research to optimize plant growth under controlled conditions.