Learn More
Medchemexpress LLC Zeaxanthin | 144-68-3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1203181MG
Encompass_Preferred
Zeaxanthin is a diet-derived xanthophyll carotenoid concentrated in the macula of the eye and reported to have antioxidant activity. The compound (CAS 144-68-3) is supplied as a research-grade analytical standard, typically provided as an orange to red powder for laboratory use.
  • Diet-derived xanthophyll carotenoid.
  • Antioxidant activity relevant to ocular research.
  • Concentrated in the macula of the eye.
  • Provided as a small research pack suitable for laboratory studies.
  • Molecular formula C40H56O2 and molecular weight 568.87 g/mol.
Catalog No. 50-002-34532