Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zeaxanthin | 144-68-3 | 10 MG
SDP

Catalog No. 5000232579
Encompass_Preferred

Zeaxanthin is a diet-derived xanthophyll carotenoid concentrated in the macula of the eye and used in research for its antioxidant properties.

  • Present in the macula and contributes to visual protection.
  • Exhibits antioxidant activity in biological systems.
  • Supplied in small milligram packages suitable for laboratory research.
  • Molecular formula C40H56O2; molecular weight 568.87 g/mol.
  • Store at -20°C and protect from light; unstable in solution, prepare fresh solutions.

Catalog No. 50-002-32579 Supplier Medchemexpress LLC Supplier No. HY12031810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zeaxanthin is a diet-derived xanthophyll carotenoid concentrated in the macula of the eye and used in research for its antioxidant properties.

  • Present in the macula and contributes to visual protection.
  • Exhibits antioxidant activity in biological systems.
  • Supplied in small milligram packages suitable for laboratory research.
  • Molecular formula C40H56O2; molecular weight 568.87 g/mol.
  • Store at -20°C and protect from light; unstable in solution, prepare fresh solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.