Learn More
Medchemexpress LLC Zeaxanthin | 144-68-3 | 95.0% | 50 MG
SDP
Catalog No. 5000231701
Encompass_Preferred
Zeaxanthin is a naturally occurring xanthophyll carotenoid and antioxidant found in the macula of the human eye. It is supplied as a research reagent for studies of retinal health, oxidative stress, and carotenoid biochemistry.
  • Acts as an antioxidant in retinal tissue.
  • Concentrated in the macula and associated with visual function.
  • Commonly used in research on age-related macular degeneration and eye health.
  • Molecular formula C40H56O2, molecular weight 568.9 g/mol.
Catalog No. 50-002-31701 Supplier Medchemexpress LLC Supplier No. HY12031850MG
Zeaxanthin is a naturally occurring xanthophyll carotenoid and antioxidant found in the macula of the human eye. It is supplied as a research reagent for studies of retinal health, oxidative stress, and carotenoid biochemistry.
  • Acts as an antioxidant in retinal tissue.
  • Concentrated in the macula and associated with visual function.
  • Commonly used in research on age-related macular degeneration and eye health.
  • Molecular formula C40H56O2, molecular weight 568.9 g/mol.