Learn More
Medchemexpress LLC Zeaxanthin | 144-68-3 | ≥97.0% | 100 MG
SDP
Catalog No. 5000231719
Encompass_Preferred
Zeaxanthin is a diet-obtained xanthophyll carotenoid found in the macula region of the eye. It exhibits antioxidant activity and is commonly used in research on retinal health and oxidative stress. The compound is unstable in solution; freshly prepare stock solutions prior to use.
  • Antioxidant properties support studies of oxidative stress.
  • Present in the macula, relevant to ocular research.
  • Lipid-soluble, suitable for studies requiring lipophilic pigments.
  • Unstable in solution; prepare fresh stock solutions.
  • Supplied as a defined chemical entity for research applications.
Catalog No. 50-002-31719 Supplier Medchemexpress LLC Supplier No. HY120318100MG
Zeaxanthin is a diet-obtained xanthophyll carotenoid found in the macula region of the eye. It exhibits antioxidant activity and is commonly used in research on retinal health and oxidative stress. The compound is unstable in solution; freshly prepare stock solutions prior to use.
  • Antioxidant properties support studies of oxidative stress.
  • Present in the macula, relevant to ocular research.
  • Lipid-soluble, suitable for studies requiring lipophilic pigments.
  • Unstable in solution; prepare fresh stock solutions.
  • Supplied as a defined chemical entity for research applications.