Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zeaxanthin | 144-68-3 | >98.0% | 5 MG
SDP

Catalog No. 5000233495
Encompass_Preferred

Zeaxanthin is a diet-obtained xanthophyll carotenoid concentrated in the macula of the eye. It exhibits antioxidant activity and is commonly used in biochemical and ocular research. Supplied as a purified reagent, it is provided in small research pack sizes and should be stored protected from light to maintain stability.

  • High purity (>98.0%) suitable for biochemical and ocular studies.
  • Molecular weight 568.87 g/mol; formula C40H56O2.
  • Available in small quantities (for example, 5 mg) for assay development and pilot studies.
  • Store at -20°C and protect from light; in solvent store at -80°C for long-term storage.
  • Intended for research use only; request certificate of analysis and safety data sheet.

Catalog No. 50-002-33495 Supplier Medchemexpress LLC Supplier No. HY1203185MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zeaxanthin is a diet-obtained xanthophyll carotenoid concentrated in the macula of the eye. It exhibits antioxidant activity and is commonly used in biochemical and ocular research. Supplied as a purified reagent, it is provided in small research pack sizes and should be stored protected from light to maintain stability.

  • High purity (>98.0%) suitable for biochemical and ocular studies.
  • Molecular weight 568.87 g/mol; formula C40H56O2.
  • Available in small quantities (for example, 5 mg) for assay development and pilot studies.
  • Store at -20°C and protect from light; in solvent store at -80°C for long-term storage.
  • Intended for research use only; request certificate of analysis and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.