Learn More
Cayman Chemical ZeaxnthIn DIpalmItate 10mg
SDP
Catalog No. 502991721
Encompass_Preferred
A beta-acid active against B subtilis and S aureus (MICs 1 and 1 2 Ug/ml respectively) and T b brucei and L m mexicana (IC50s 0 9 and 4 7 Ug/ml respectively) scavenges DPPH radicals in a cell-free assay and inhibits lipid peroxidation in rat brain homogenates (IC50s 25 and 39 UM respectively) reduces proliferation cell migration and capillary tube formation in HUVECs at 2 5 to 50 Ug/ml lupulone (40 Ug/ml) activates the extrinsic apoptotic death pathway in SW480 and SW620 colon cancer cells
Catalog No. 50-299-1721 Supplier Cayman Chemical Supplier No. 3170310 MG
A beta-acid active against B subtilis and S aureus (MICs 1 and 1 2 Ug/ml respectively) and T b brucei and L m mexicana (IC50s 0 9 and 4 7 Ug/ml respectively) scavenges DPPH radicals in a cell-free assay and inhibits lipid peroxidation in rat brain homogenates (IC50s 25 and 39 UM respectively) reduces proliferation cell migration and capillary tube formation in HUVECs at 2 5 to 50 Ug/ml lupulone (40 Ug/ml) activates the extrinsic apoptotic death pathway in SW480 and SW620 colon cancer cells