Learn More
Zebra Technologies Corporation Label, Polyester, 2x0.25in (50.8x6.4mm); TT, Z-Ultimate 2000T White, Value Coated, Permanent Adhesive, 1in (25.4mm) core, 2500/roll, 1/box, Plain
Catalog No. NC2828646
Encompass_Preferred
This item has a minimum order quantity of 1 per supplier requirements
Label, Polyester, 2x0.25in (50.8x6.4mm); TT, Z-Ultimate 2000T White, Value Coated, Permanent Adhesive, 1in (25.4mm) core, 2500/roll, 1/box, Plain
Catalog No. NC2828646 Supplier Zebra Technologies Corporation Supplier No. 10023033
This item has a minimum order quantity of 1 per supplier requirements
Label, Polyester, 2x0.25in (50.8x6.4mm); TT, Z-Ultimate 2000T White, Value Coated, Permanent Adhesive, 1in (25.4mm) core, 2500/roll, 1/box, Plain