Learn More
BARCODESINC ZEBRA BARCODE LABEL PRINTER
Catalog No. NC2208990
Encompass
Barcode Label Printer, Zebra printer ZT411 Zebra ZT411 - Industrial Label Printer, Zebra ZT411, Thermal transfer, 203 dpi, 14 ips, Ethernet/Serial/USB/USB Host/Bluetooth, 4-inch Print width, Includes US Power cord, EZPL.A smart choice for carrying out high volume label printing in multiple areas with reduced cost of ownershipEnjoy the optimal performance in today's on-demand labeling applicationsSave time and money for your business by using one printer for all your employees
This item is not returnable. View return policy
Catalog No. NC2208990 Supplier BARCODESINC Supplier No. ZT41142T010000Z
Barcode Label Printer, Zebra printer ZT411 Zebra ZT411 - Industrial Label Printer, Zebra ZT411, Thermal transfer, 203 dpi, 14 ips, Ethernet/Serial/USB/USB Host/Bluetooth, 4-inch Print width, Includes US Power cord, EZPL.A smart choice for carrying out high volume label printing in multiple areas with reduced cost of ownershipEnjoy the optimal performance in today's on-demand labeling applicationsSave time and money for your business by using one printer for all your employees