Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BARCODESINC ZEBRA DS2208 - GENERAL PURPOSE

Catalog No. NC3073929
Encompass

  • Efficient and reliable barcode scanning for diverse environments.
  • Capable of scanning 1D and 2D barcodes, including QR codes and Data Matrix.
  • Omnidirectional scanning eliminates need for precise alignment.
  • Plug-and-play USB connectivity for easy integration with existing POS systems.
  • Durable design withstands multiple 5.0 ft drops to concrete.
  • Advanced scanning technology with Zebra's PRZM software algorithms.
  • Max working distance of 14.5 in.
  • Scan speed of 30 in/sec for quick transactions.
  • Supported host interfaces include USB, RS232, and Keyboard Wedge.
  • Light source wavelength of 624 nm.
  • Includes shielded USB cable of 7 ft length.

Catalog No. NC3073929 Supplier BARCODESINC Supplier No. DS2208SR7U2100AZW
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Efficient and reliable barcode scanning for diverse environments.
  • Capable of scanning 1D and 2D barcodes, including QR codes and Data Matrix.
  • Omnidirectional scanning eliminates need for precise alignment.
  • Plug-and-play USB connectivity for easy integration with existing POS systems.
  • Durable design withstands multiple 5.0 ft drops to concrete.
  • Advanced scanning technology with Zebra's PRZM software algorithms.
  • Max working distance of 14.5 in.
  • Scan speed of 30 in/sec for quick transactions.
  • Supported host interfaces include USB, RS232, and Keyboard Wedge.
  • Light source wavelength of 624 nm.
  • Includes shielded USB cable of 7 ft length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.