Learn More
BARCODESINC ZEBRA DS8178 SCANNER
Catalog No. NC2582161
Encompass_Preferred
  • Cordless, standard-range handheld area imager.
  • Rapid and accurate scanning of 1D and 2D barcodes, even on damaged or poorly printed surfaces.
  • 800 MHz microprocessor with high-resolution megapixel sensor for exceptional performance.
  • Scan range of up to 24 in (61 cm), allowing effective data capture from hard-to-reach items.
  • Flexible power options with swappable PowerPrecision+ batteries or PowerCap capacitors.
  • Battery life exceeds 3 days of continuous scanning.
  • Robust design withstands drops from 6 ft (1.8 m) onto concrete.
  • IP52 or IP42 environmental sealing for reliable operation in challenging conditions.
  • Bluetooth Version 4.0 connectivity with 33 ft (10 m) range.
  • Comes with tools for barcode issue identification and proactive power monitoring.
Catalog No. NC2582161 Supplier BARCODESINC Supplier No. DS8178SR0F007ZZWW
  • Cordless, standard-range handheld area imager.
  • Rapid and accurate scanning of 1D and 2D barcodes, even on damaged or poorly printed surfaces.
  • 800 MHz microprocessor with high-resolution megapixel sensor for exceptional performance.
  • Scan range of up to 24 in (61 cm), allowing effective data capture from hard-to-reach items.
  • Flexible power options with swappable PowerPrecision+ batteries or PowerCap capacitors.
  • Battery life exceeds 3 days of continuous scanning.
  • Robust design withstands drops from 6 ft (1.8 m) onto concrete.
  • IP52 or IP42 environmental sealing for reliable operation in challenging conditions.
  • Bluetooth Version 4.0 connectivity with 33 ft (10 m) range.
  • Comes with tools for barcode issue identification and proactive power monitoring.