Learn More
BARCODESINC ZEBRA ZD421 PRINTER,203DPI
Catalog No. NC2051702
Encompass_Preferred
Desktop Printer, Zebra ZD421, Thermal Transfer,203 dpi, 4ips, USB/USB Host/Ethernet/Bluetooth,ZPL/EPL, 4-inch, Standard Tear
Catalog No. NC2051702 Supplier BARCODESINC Supplier No. ZD4A042301E00EZ
Desktop Printer, Zebra ZD421, Thermal Transfer,203 dpi, 4ips, USB/USB Host/Ethernet/Bluetooth,ZPL/EPL, 4-inch, Standard Tear