Learn More
Uline Zebra ZD421T Dual Barcode Printer, Desktop
Catalog No. NC2330827
Encompass
Zebra ZD421T desktop barcode printer with dual printheads for printing 2 labels or tags simultaneously; prints 2D barcodes including QR; USB / Bluetooth LE interface, 8 x 10 1/2 x 7 1/2""""
Catalog No. NC2330827 Supplier Uline Supplier No. H9582
Zebra ZD421T desktop barcode printer with dual printheads for printing 2 labels or tags simultaneously; prints 2D barcodes including QR; USB / Bluetooth LE interface, 8 x 10 1/2 x 7 1/2""""