Learn More
GA International Zebra ZT231 Thermal-Transfer/Direct Thermal Printer
Catalog No. 5000819169
Encompass
The ZT231 offers an optimized design and smaller footprint that takes up less physical space than other similar printers, with a durable design to minimize maintenance. Built for light industrial and commercial environments, the ZT231 printer is ideal for large volume printing, able to print 300 dpi resolution text and barcodes at up to 8″/second. The ZT231 printer has an easy-to-use display and can print in both thermal-transfer and direct thermal modes, great for a variety of labeling applications. The printer also provides various connectivity options, such as USB, parallel, and Ethernet. Printing kits are available on demand. Contact our technical support team for further information.
Catalog No. 50-008-19169 Supplier GA International Supplier No. ZT231300
The ZT231 offers an optimized design and smaller footprint that takes up less physical space than other similar printers, with a durable design to minimize maintenance. Built for light industrial and commercial environments, the ZT231 printer is ideal for large volume printing, able to print 300 dpi resolution text and barcodes at up to 8″/second. The ZT231 printer has an easy-to-use display and can print in both thermal-transfer and direct thermal modes, great for a variety of labeling applications. The printer also provides various connectivity options, such as USB, parallel, and Ethernet. Printing kits are available on demand. Contact our technical support team for further information.