Learn More
Biomatik Corporation Zebrafish Gonadotropin Releasing Hormone (GnRH) ELISA Kit
Supplier:  Biomatik Corporation EKU11936
Encompass_Preferred
Zebrafish Gonadotropin Releasing Hormone (GnRH) ELISA Kit for detecting GnRH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Zebrafish. Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.39pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: F1QW13. Gene ID: 360141. Target Synonyms: GNRH1; GRH; LNRH; LHRH; LH-RH I; Luliberin; Luteinizing-Hormone Releasing Hormone; Progonadoliberin-1; Gonadoliberin I; Gonadorelin; GnRH-associated peptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7376