Learn More
Kingfisher Biotech Inc Canine Neurophysin-1 ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1241Z004
Encompass_Preferred
The Canine Neurophysin-1 ELISA contains capture antibody, standard, and detection antibody for development of a Canine Neurophysin-1 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA.
Catalog No. 50-228-5384