Learn More
Cayman Chemical Zebrafish Androgen Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502947818
Encompass_Preferred
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spinand-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.
Catalog No. 50-294-7818 Supplier Cayman Chemical Supplier No. 3257896 WELL
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spinand-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.