Learn More
Cayman Chemical ZebrafIsh ndrogen RECPTR R
SDP
Catalog No. 502947682
Encompass_Preferred
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spinand-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.
Catalog No. 50-294-7682 Supplier Cayman Chemical Supplier No. 325783X32 WELL
Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spinand-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.