Learn More
Cayman Chemical Zebrafish Peroxisome Proliferator-Activated Receptor Gamma Reporter Assay System, 96 Well
SDP
Catalog No. 502949410
Encompass_Preferred
zPPARγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. Z00101-32, Z00101]
Catalog No. 50-294-9410 Supplier Cayman Chemical Supplier No. 3257596 WELL
zPPARγ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. Z00101-32, Z00101]